Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Nitroacetophenone, 95%, Thermo Scientific™

Catalog No. AC128320250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC128320250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC128320250 Supplier Thermo Scientific Chemicals Supplier No. 128320250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-59-3
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007144
InChI Key SUGXZLKUDLDTKX-UHFFFAOYSA-N
Synonym 2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
PubChem CID 11346
IUPAC Name 1-(2-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 28.0°C to 30.0°C
Boiling Point 159.0°C (16.0 mmHg)
Flash Point >112°C
Refractive Index 1.5500 to 1.5520
Linear Formula O2NC6H4COCH3
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
Formula Weight 165.15
Chemical Name or Material o-Nitroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.