Learn More
Medchemexpress LLC o-nitrophenyl β-D-galactopyranoside-6-phosphate (cyclohexylammonium) | 25405-62-3 | 99.0% | 480.40 | C18H29N2O11P | 10 MG
SDP
Catalog No. 5000283913
Encompass_Preferred
o-Nitrophenyl β-D-galactopyranoside-6-phosphate (cyclohexylammonium) is a chromogenic substrate for detection and quantification of β-galactosidase activity in biochemical assays. The phosphate salt form improves solubility and handling for assay preparation and supports reproducible colorimetric readouts.
  • Chromogenic readout produces a measurable colored product upon enzyme hydrolysis.
  • Suitable for quantitative β-galactosidase assays and activity screening.
  • Phosphate salt form improves solubility and assay preparation.
  • High purity supports consistent and reliable assay performance.
  • Available in small research-scale quantities for assay optimization.
Catalog No. 50-002-83913 Supplier Medchemexpress LLC Supplier No. HYW58777910MG
o-Nitrophenyl β-D-galactopyranoside-6-phosphate (cyclohexylammonium) is a chromogenic substrate for detection and quantification of β-galactosidase activity in biochemical assays. The phosphate salt form improves solubility and handling for assay preparation and supports reproducible colorimetric readouts.
  • Chromogenic readout produces a measurable colored product upon enzyme hydrolysis.
  • Suitable for quantitative β-galactosidase assays and activity screening.
  • Phosphate salt form improves solubility and assay preparation.
  • High purity supports consistent and reliable assay performance.
  • Available in small research-scale quantities for assay optimization.