Learn More
Medchemexpress LLC o-Nitrophenyl β-D-galactopyranoside-6-phosphate cyclohexylammonium | 25405-62-3 | MFCD00083446 | 98.0% | 480.4 | C18H29N2O11P | 5 MG
SDP
Catalog No. 5000280475
Encompass_Preferred
o-Nitrophenyl β-D-galactopyranoside-6-phosphate cyclohexylammonium is a chromogenic substrate used to detect and quantify β-galactosidase activity. Upon enzymatic hydrolysis it releases a nitrophenyl chromophore that produces a measurable color change suitable for rapid colorimetric or spectrophotometric assays in biochemical and enzymology applications.
  • Chromogenic substrate for β-galactosidase detection.
  • Suitable for colorimetric and spectrophotometric assays.
  • Enables rapid, quantitative measurement of enzyme activity.
  • Water-soluble cyclohexylammonium salt for assay compatibility.
  • Supplied in small research-scale quantities for assay development.
Catalog No. 50-002-80475 Supplier Medchemexpress LLC Supplier No. HYW5877795MG
o-Nitrophenyl β-D-galactopyranoside-6-phosphate cyclohexylammonium is a chromogenic substrate used to detect and quantify β-galactosidase activity. Upon enzymatic hydrolysis it releases a nitrophenyl chromophore that produces a measurable color change suitable for rapid colorimetric or spectrophotometric assays in biochemical and enzymology applications.
  • Chromogenic substrate for β-galactosidase detection.
  • Suitable for colorimetric and spectrophotometric assays.
  • Enables rapid, quantitative measurement of enzyme activity.
  • Water-soluble cyclohexylammonium salt for assay compatibility.
  • Supplied in small research-scale quantities for assay development.