Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Nitrophenyl-β-D-galactopyranoside, MP Biomedicals™

Catalog No. ICN10247325 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10247325 25 g
ICN10247301 1 g
ICN10247305 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10247325 Supplier MP Biomedicals, Inc Supplier No. 0210247325
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

o-Nitrophenyl-β-D-galactoside is a cell culture component for molecular genetics

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-07-3
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00063255
InChI Key KUWPCJHYPSUOFW-YBXAARCKSA-N
Synonym 2-nitrophenyl-beta-d-galactopyranoside, onpg, 2-nitrophenyl beta-d-galactopyranoside, 2-nitrophenyl beta-d-galactoside, o-nitrophenyl beta-d-galactopyranoside, o-nitrophenyl-beta-d-galactoside, 1-o-o-nitrophenyl-beta-d-galactopyranose, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy oxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2-nitrophenoxy tetrahydro-2h-pyran-3,4,5-triol, 2-nitrophenyl b-d-galactopyranoside
PubChem CID 96647
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(2-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

Specifications

Color White
Quantity 25 g
Formula Weight 301.3
Percent Purity 99%
Grade Not Available
Physical Form Solid
Chemical Name or Material o-Nitrophenyl-β-D-galactopyranoside
EINECSNumber 206-716-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.