Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ O-Nitrophenyl-β-D-Galactopyranoside, MP Biomedicals™

Catalog No. mp021024735 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021024735 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021024735 Supplier MP Biomedicals™ Supplier No. 02102473.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Useful in the detection of lacZ activity. O-Nitrophenyl-β-D-Galactopyranoside, MP Biomedicals is a colorimetric substrate for b-galactosidase and a cell culture component for molecular genetics. ONPG reactions are rapid and occur within a 24-hour per

This product is hydrolyzed to O-Nitrophenol by the enzyme. O-Nitrophenol is yellow in alkaline solution and can be quantitated at 410-420nm2.

  • β-galactosidase breaks down lactose into galactose and glucose
  • β-galactosidase is not lactose specific and can act on simple galactosides
  • 2-Nitrophenyl β-D-galactopyranoside hydrolysis results in the release of galactose and a yellow chromogenic compound
  • The test substrate does not depend on an induced or constitutive permease enzyme to enter the cell
TRUSTED_SUSTAINABILITY

Specifications

Form Solid
For Use With (Application) o-Nitrophenyl-β-D-galactoside is a cell culture component for molecular genetics. It is chromogenic substrate for b-galactosidase and hence is useful in the detection of lacZ activity.
Quantity 500mg
Format White Solid
Molecular Weight (g/mol) 301.3
pH −66° ± 7° (c=1, H2O)
Storage Requirements 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.