Learn More
Medchemexpress LLC O-Nornuciferine | 3153-55-7 | 10 MG
SDP
Catalog No. 5000417585
Encompass_Preferred
O-Nornuciferine, an aporphine-type alkaloid derived from lotus leaf, is recognized as a potent inhibitor of the hERG channel.
  • It is an endogenous metabolite.
  • It shows an IC50 of 7.91 μM in in vitro hERG blockage experiments using HEK293 cells.
  • At 100 μM, it demonstrates 47% hERG channel inhibition in Xenopus oocytes.
  • The purity of the product is 99.85%.
Catalog No. 50-004-17585 Supplier Medchemexpress LLC Supplier No. HYN751110MG
O-Nornuciferine, an aporphine-type alkaloid derived from lotus leaf, is recognized as a potent inhibitor of the hERG channel.
  • It is an endogenous metabolite.
  • It shows an IC50 of 7.91 μM in in vitro hERG blockage experiments using HEK293 cells.
  • At 100 μM, it demonstrates 47% hERG channel inhibition in Xenopus oocytes.
  • The purity of the product is 99.85%.