Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O-Nornuciferine | 3153-55-7 | 5 MG
SDP

Catalog No. 5000419394
Encompass_Preferred

O-Nornuciferine is an aporphine-type alkaloid derived from lotus leaf. It acts as a potent hERG channel inhibitor, exhibiting an IC50 of 7.91 μM in vitro hERG blockages in HEK293 cells. This compound is intended for research use only.

  • Potent hERG channel inhibitor
  • 99.85% purity
  • White to off-white solid appearance
  • Store at 4°C, sealed, away from moisture and light
  • Soluble in DMSO at 50 mg/mL

Catalog No. 50-004-19394 Supplier Medchemexpress LLC Supplier No. HYN75115MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-Nornuciferine is an aporphine-type alkaloid derived from lotus leaf. It acts as a potent hERG channel inhibitor, exhibiting an IC50 of 7.91 μM in vitro hERG blockages in HEK293 cells. This compound is intended for research use only.

  • Potent hERG channel inhibitor
  • 99.85% purity
  • White to off-white solid appearance
  • Store at 4°C, sealed, away from moisture and light
  • Soluble in DMSO at 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.