Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich O,O'-Bis(2-aminopropyl) polypropylene glycol-block-polyethylene glycol-block-polypropylene glycol1

Catalog No. 14527500MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-527-500MLF 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-527-500MLF Supplier Sigma Aldrich Supplier No. 14527500MLF
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Jeffamine(R) ED-900 is an amine-terminated poly(ethylene glycol)/poly(propylene glycol) copolymer. It exists as a low viscosity and low vapor pressure liquid.{4}Jeffamine(R) ED-900 may be used to fabricate thin electrolyte films in diureasil matrix.{5} Succinyl chloride polymerized with ED-900 may be used as a drug carrier.{6}Jeffamine is a registered trademark of Huntsman Petrochemical, LLC

Specifications

CAS 65605-36-9
Linear Formula CH3CH(NH2)CH2[OCH(CH3)CH2]l(OCH2CH2)m[OCH2CH(CH3)]nNH2
MDL Number MFCD00147794
Quantity 500 mL
Synonym Jeffamine™ ED-900
Molecular Weight (g/mol) 900
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.