Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O,O-Dimethyl-O(2,2-dichlorovinyl) Phosphate, MP Biomedicals

Catalog No. ICN203063 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN203063 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN203063 Supplier MP Biomedicals, Inc Supplier No. 0520306310

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 62-73-7
Molecular Formula C4H7Cl2O4P
Molecular Weight (g/mol) 220.97
MDL Number MFCD00036123
InChI Key OEBRKCOSUFCWJD-UHFFFAOYSA-N
Synonym dichlorvos, ddvp, dichlorophos, divipan, vapona, 2,2-dichlorovinyl dimethyl phosphate, chlorvinphos, brevinyl, canogard, dichlorphos
PubChem CID 3039
ChEBI CHEBI:34690
IUPAC Name 2,2-dichloroethenyl dimethyl phosphate
SMILES COP(=O)(OC)OC=C(Cl)Cl

Specifications

Quantity 10 g
Formula Weight 220.9
Chemical Name or Material O, O-Dimethyl-O(2, 2-Dichlorovinyl) Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.