Learn More
Medchemexpress LLC O,O'-[Oxybis(2,1-ethanediyloxy-2,1-ethanediyl)]bis[hydroxylamine] | 98627-70-4 | 98.0% | C8H20N2O5 | 100 MG
SDP
Catalog No. 5000356642
Encompass_Preferred
This product, Bis-aminooxy-PEG3, is a PEG-based PROTAC linker. It has a molecular weight of 224.25 and appears as a colorless to light yellow liquid.
  • Consistent with structure based on ¹H NMR spectrum
  • Consistent with structure based on MS
Catalog No. 50-003-56642 Supplier Medchemexpress LLC Supplier No. HY140409100MG
This product, Bis-aminooxy-PEG3, is a PEG-based PROTAC linker. It has a molecular weight of 224.25 and appears as a colorless to light yellow liquid.
  • Consistent with structure based on ¹H NMR spectrum
  • Consistent with structure based on MS