Learn More
Medchemexpress LLC o,p'-DDE (2,4'-DDE) | 3424-82-6 | >95.0% | 5 MG
SDP
Catalog No. 5000231523
Encompass_Preferred
o,p'-DDE (2,4'-DDE) is an organochlorine pesticide metabolite and degradation product of DDT used as an analytical reference standard in toxicology and environmental monitoring. It is monitored for persistence and potential endocrine activity.
  • Used as an analytical reference standard for environmental and toxicology assays.
  • Shows documented interaction with estrogen receptors in vitro.
  • Relevant for studies of persistent organic pollutants and bioaccumulation.
  • Available in small, defined quantities suitable for laboratory research.
Catalog No. 50-002-31523 Supplier Medchemexpress LLC Supplier No. HY1217795MG
o,p'-DDE (2,4'-DDE) is an organochlorine pesticide metabolite and degradation product of DDT used as an analytical reference standard in toxicology and environmental monitoring. It is monitored for persistence and potential endocrine activity.
  • Used as an analytical reference standard for environmental and toxicology assays.
  • Shows documented interaction with estrogen receptors in vitro.
  • Relevant for studies of persistent organic pollutants and bioaccumulation.
  • Available in small, defined quantities suitable for laboratory research.