Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC o,p'-DDE (2,4'-DDE) | 3424-82-6 | >=95.0% | 50 MG
SDP

Catalog No. 5000232837
Encompass_Preferred

o,p'-DDE (2,4'-DDE) is an organochlorine pesticide metabolite and degradation product of DDT commonly used as an analytical standard and in toxicological and environmental research.

  • Metabolite of DDT commonly used as an analytical standard
  • CAS number 3424-82-6
  • Molecular formula C14H8Cl4, molecular weight 318.03 g/mol
  • Provided as a small solid package suitable for laboratory use
  • Purity not specified on product page; check Certificate of Analysis for lot-specific purity

Catalog No. 50-002-32837 Supplier Medchemexpress LLC Supplier No. HY12177950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

o,p'-DDE (2,4'-DDE) is an organochlorine pesticide metabolite and degradation product of DDT commonly used as an analytical standard and in toxicological and environmental research.

  • Metabolite of DDT commonly used as an analytical standard
  • CAS number 3424-82-6
  • Molecular formula C14H8Cl4, molecular weight 318.03 g/mol
  • Provided as a small solid package suitable for laboratory use
  • Purity not specified on product page; check Certificate of Analysis for lot-specific purity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.