Learn More
Medchemexpress LLC o,p'-DDE (2,4'-DDE) | 3424-82-6 | 98.0% | 1 MG
SDP
Catalog No. 5000234825
Encompass_Preferred
o,p'-DDE (2,4'-DDE) is an organochlorine pesticide metabolite and degradation product of DDT used as an analytical reference in environmental and toxicology research. It is a persistent organic pollutant with documented endocrine-disrupting activity; handle and dispose according to institutional and regulatory safety guidance.
  • Analytical reference standard for environmental and toxicology studies.
  • Chemical formula C14H8Cl4; molecular weight 318.03 g/mol.
  • Available in milligram pack sizes suitable for trace-level analyses.
  • Useful for method development, calibration, and toxicological assays.
  • Persistent organic pollutant; requires appropriate handling and disposal.
Catalog No. 50-002-34825 Supplier Medchemexpress LLC Supplier No. HY1217791MG
o,p'-DDE (2,4'-DDE) is an organochlorine pesticide metabolite and degradation product of DDT used as an analytical reference in environmental and toxicology research. It is a persistent organic pollutant with documented endocrine-disrupting activity; handle and dispose according to institutional and regulatory safety guidance.
  • Analytical reference standard for environmental and toxicology studies.
  • Chemical formula C14H8Cl4; molecular weight 318.03 g/mol.
  • Available in milligram pack sizes suitable for trace-level analyses.
  • Useful for method development, calibration, and toxicological assays.
  • Persistent organic pollutant; requires appropriate handling and disposal.