Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC o,p'-DDE (2,4'-dichlorodiphenyldichloroethylene) | 3424-82-6 | 25 MG
SDP

Catalog No. 5000231524
Encompass_Preferred

o,p'-DDE (2,4'-DDE) is an organochlorine pesticide metabolite and degradation product of DDT supplied as an analytical reference standard for environmental and toxicology research. It is provided in small vial quantities suitable for laboratory analytical workflows.

  • Used as an analytical standard for environmental and toxicology analysis.
  • Metabolite and degradation product of DDT.
  • Chemical formula C14H8Cl4.
  • Molecular weight 318.03 g/mol.
  • Available in small vial sizes including 25 mg for lab-scale analyses.
  • Refer to the product datasheet for exact purity and handling instructions.

Catalog No. 50-002-31524 Supplier Medchemexpress LLC Supplier No. HY12177925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

o,p'-DDE (2,4'-DDE) is an organochlorine pesticide metabolite and degradation product of DDT supplied as an analytical reference standard for environmental and toxicology research. It is provided in small vial quantities suitable for laboratory analytical workflows.

  • Used as an analytical standard for environmental and toxicology analysis.
  • Metabolite and degradation product of DDT.
  • Chemical formula C14H8Cl4.
  • Molecular weight 318.03 g/mol.
  • Available in small vial sizes including 25 mg for lab-scale analyses.
  • Refer to the product datasheet for exact purity and handling instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.