Learn More
Selleck Chemical LLC o-Phenanthroline
SDP
Catalog No. 5000002737
Encompass_Preferred
o-Phenanthroline (1 10-Phenanthroline phen) a hypoxia-mimicking agent is a non-specific matrix metalloproteinase (MMP) inhibitor with IC50 of 110 5 M for Collagenase O-phenanthroline reduces the expression of MMP3 and MMP13 mRNA levels during chondrogenic differentiation of human chondrocytes (hChs) as well as after TNF /IL-1 exposure in an explant model o-Phenanthroline is also a metal chelator which prevents the induction of chromosomal aberrations in streptozotocin-treated cells and forms a red chelate with Fe2 that absorbs maximally at 510 nm
Catalog No. 50-000-02737 Supplier Selleck Chemical LLC Supplier No. S6830100MG
o-Phenanthroline (1 10-Phenanthroline phen) a hypoxia-mimicking agent is a non-specific matrix metalloproteinase (MMP) inhibitor with IC50 of 110 5 M for Collagenase O-phenanthroline reduces the expression of MMP3 and MMP13 mRNA levels during chondrogenic differentiation of human chondrocytes (hChs) as well as after TNF /IL-1 exposure in an explant model o-Phenanthroline is also a metal chelator which prevents the induction of chromosomal aberrations in streptozotocin-treated cells and forms a red chelate with Fe2 that absorbs maximally at 510 nm