Learn More
Medchemexpress LLC o-Phenanthroline | 66-71-7 | 180.21 | 1 ML
SDP
Catalog No. 5000448772
Encompass_Preferred
o-Phenanthroline (1,10-Phenanthroline) is a metal chelator that prevents the induction of chromosomal aberrations in streptozotocin-treated cells. It forms a red chelate with Fe2+ that absorbs maximally at 510 nm and also acts as an MMP inhibitor.
  • Acts as a metal chelator
  • Prevents induction of chromosomal aberrations
  • Forms a red chelate with Fe2+
  • Functions as an MMP inhibitor
  • Demonstrates diverse solubility in various solvents
  • Exhibits broad biological activity across multiple cell lines
Catalog No. 50-004-48772 Supplier Medchemexpress LLC Supplier No. HYW00454410MM/1ML
Add to Cart
Add to Cart
o-Phenanthroline (1,10-Phenanthroline) is a metal chelator that prevents the induction of chromosomal aberrations in streptozotocin-treated cells. It forms a red chelate with Fe2+ that absorbs maximally at 510 nm and also acts as an MMP inhibitor.
  • Acts as a metal chelator
  • Prevents induction of chromosomal aberrations
  • Forms a red chelate with Fe2+
  • Functions as an MMP inhibitor
  • Demonstrates diverse solubility in various solvents
  • Exhibits broad biological activity across multiple cell lines