Learn More
Apexbio Technology LLC o-Phenanthroline monohydrate 5144-89-8 100mg
SDP
Catalog No. 5000095099
Encompass_Preferred
o-Phenanthroline monohydrate (CAS 5144-89-8) is a small-molecule inhibitor targeting matrix metalloproteinases (MMPs) It is designed to chelate metal ions thereby inhibiting the catalytic activity of MMPs and modulating related biological pathways o-Phenanthroline monohydrate exerts its biological activity primarily through metal ion chelation particularly by complexing with ferrous ions to form a chromophoric complex It demonstrates inhibitory activity in research involving MMPs and interference with chemically induced chromosomal aberrations Based on these pharmacological properties o-Phenanthroline monohydrate holds research potential in studies of cell migration tumor invasion signaling pathways and chromosome aberration modulation
Catalog No. 50-000-95099 Supplier Apexbio Technology LLC Supplier No. A4741100
o-Phenanthroline monohydrate (CAS 5144-89-8) is a small-molecule inhibitor targeting matrix metalloproteinases (MMPs) It is designed to chelate metal ions thereby inhibiting the catalytic activity of MMPs and modulating related biological pathways o-Phenanthroline monohydrate exerts its biological activity primarily through metal ion chelation particularly by complexing with ferrous ions to form a chromophoric complex It demonstrates inhibitory activity in research involving MMPs and interference with chemically induced chromosomal aberrations Based on these pharmacological properties o-Phenanthroline monohydrate holds research potential in studies of cell migration tumor invasion signaling pathways and chromosome aberration modulation