Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC O-phenylenediacetic acid | 7500-53-0 | 194.18 | 25 G
SDP

Catalog No. 5000448251
Encompass_Preferred

O-phenylenediacetic acid, also known as 1,2-Benzenediacetic acid, is a biochemical reagent. It is intended for research use in life science applications as a biological material or organic compound.

  • High purity: 99.96%
  • Molecular weight: 194.18
  • Formula: C10H10O4
  • Appearance: Off-white to light brown solid
  • Long-term stability when stored at room temperature
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-48251 Supplier Medchemexpress LLC Supplier No. 25GHYW008103
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

O-phenylenediacetic acid, also known as 1,2-Benzenediacetic acid, is a biochemical reagent. It is intended for research use in life science applications as a biological material or organic compound.

  • High purity: 99.96%
  • Molecular weight: 194.18
  • Formula: C10H10O4
  • Appearance: Off-white to light brown solid
  • Long-term stability when stored at room temperature
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.