Learn More
Alkali Scientific o Phenylenediamine, 50 Grams
SDP
Catalog No. 5000661811
Encompass_Preferred
O Phenylenediamine, available in a 50g package, is an aromatic compound commonly used in laboratory research and chemical synthesis. It is often employed as a reagent in the determination of peroxide levels, the development of colorimetric assays, and in the synthesis of certain dyes and polymers. O Phenylenediamine is also used as a precursor in the synthesis of various organic compounds, including pharmaceuticals and agrochemicals. This high reactivity compound is particularly useful in analytical chemistry, and its versatility makes it an essential tool in chemical and biochemical laboratories for a wide range of applications, including enzyme assays and staining procedures.
Catalog No. 50-006-61811 Supplier Alkali Scientific Supplier No. OPH001CP
O Phenylenediamine, available in a 50g package, is an aromatic compound commonly used in laboratory research and chemical synthesis. It is often employed as a reagent in the determination of peroxide levels, the development of colorimetric assays, and in the synthesis of certain dyes and polymers. O Phenylenediamine is also used as a precursor in the synthesis of various organic compounds, including pharmaceuticals and agrochemicals. This high reactivity compound is particularly useful in analytical chemistry, and its versatility makes it an essential tool in chemical and biochemical laboratories for a wide range of applications, including enzyme assays and staining procedures.