Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phenylphenol, Na Salt,MP Biomedicals

Catalog No. ICN225557 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225557 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225557 Supplier MP Biomedicals, Inc Supplier No. 0522555780

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 132-27-4
Molecular Formula C12H9NaO
Molecular Weight (g/mol) 192.193
InChI Key KSQXVLVXUFHGJQ-UHFFFAOYSA-M
Synonym sodium 1,1'-biphenyl-2-olate, sodium 2-biphenylate, sodium o-phenylphenate, 2-phenylphenol sodium salt, natriphene, orphenol, bactrol, o-phenylphenol sodium, sodium o-phenylphenol, dowicide a
PubChem CID 23675735
ChEBI CHEBI:82371
IUPAC Name sodium;2-phenylphenolate
SMILES C1=CC=C(C=C1)C2=CC=CC=C2[O-].[Na+]

Specifications

Quantity 100 g
Formula Weight 192.18
Chemical Name or Material o-PhenylPhenol, Na Salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.