Learn More
Medchemexpress LLC HY-15129 100mg Medchemexpress, O-Phospho-L-serine CAS:407-41-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15129100MG
Encompass_Preferred
Medchemexpress, HY-15129 100mg O-Phospho-L-serine CAS:407-41-0 O-Phospho-L-serine is the immediate precursor to L-serine in the serine synthesis pathway, and an agonist at the group III mGluR receptors (mGluR4, mGluR6, mGluR7, and mGluR8); O-Phospho-L-serine also acts as a weak antagonist for mGluR1 and a potent antagonist for mGluR2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1683