Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-PHOSPHO-L-SERINE, MP Biomedicals™

Catalog No. MP522549010 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP522549010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP522549010 Supplier MP Biomedicals, Inc Supplier No. 0522549010

missing translation for 'validMainframeMsg'

O-PHOSPHO-L-SERINE

  • Group III metabotropic glutamate receptor agonist (mGluR4, mGluR7, and mGluR8).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-41-0
Molecular Formula C3H8NO6P
Molecular Weight (g/mol) 185.07
MDL Number MFCD00065935
InChI Key BZQFBWGGLXLEPQ-REOHCLBHSA-N
Synonym L-2-Amino-3-hydroxypropanoic acid 3-phosphate, L-SOP, L-Serine O-phosphate

Specifications

Melting Point 190°C (literature)
Beilstein 1726826
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.