Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Phospho-L-Threonine, MP Biomedicals™

Catalog No. MP215624294 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP215624294 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215624294 Supplier MP Biomedicals, Inc Supplier No. 0215624294

Derived from the α-amino acid threonine. O-Phospho-L-Threonine, MP Biomedicals is a phosphoamino acid that is the phosphorylated ester of threonine.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1114-81-4
Molecular Formula C4H10NO6P
MDL Number MFCD00069578,MFCD00135662
Quantity 5 g
Synonym phosphothreonine, o-phospho-l-threonine, o-phosphothreonine, l-threonine phosphate, threoninium dihydrogen phosphate, l-threonine o-phosphate, h-thr po3h2-oh, l-threonine o-3-phosphate, o3-phosphothreonine, 2s,3r-2-amino-3-phosphonooxy butanoic acid
InChI Key USRGIUJOYOXOQJ-UHFFFAOYNA-N
SMILES CC(OP(O)(O)=O)C(N)C(O)=O
IUPAC Name 2-amino-3-(phosphonooxy)butanoic acid
Molecular Weight (g/mol) 199.10
PubChem CID 3246323
ChEBI CHEBI:37525
Formula Weight 199.1
Physical Form Powder
Show More Show Less
Recommended Storage −20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.