Learn More
Bioworld o-Phosphoric Acid Solution 10% v/v (1+9), 4 L
Catalog No. 50255551
Encompass_Preferred
Synonym: Orthophosphoric Acid; Phosphoric AcidUseful for HPLC10% v/v (1+9) solution in H2OAppearance: LiquidDensity : 1.335 g/mL at 20 °C
Catalog No. 50-255-551 Supplier Bioworld Supplier No. 402201213
Synonym: Orthophosphoric Acid; Phosphoric AcidUseful for HPLC10% v/v (1+9) solution in H2OAppearance: LiquidDensity : 1.335 g/mL at 20 °C