Learn More
Apexbio Technology LLC O-Phosphorylethanolamine 1071-23-4 500mg
SDP
Catalog No. 5000088787
Encompass_Preferred
O-Phosphorylethanolamine (CAS 1071-23-4) is an endogenous phospholipid metabolite involved in the biosynthesis of phosphatidylethanolamine a major component of the cytoplasmic leaflet of cellular membranes It acts as a precursor in the CDP-ethanolamine pathway contributing to membrane integrity and dynamic cellular functions such as membrane fusion autophagy and apoptosis Research indicates that reduced levels of O-phosphorylethanolamine are associated with increased neuronal loss in neurodegenerative disorders including Alzheimer s Huntington s and Parkinson s diseases This molecule is therefore of interest in studies of phospholipid metabolism and neurodegeneration
Catalog No. 50-000-88787 Supplier Apexbio Technology LLC Supplier No. B7908500
O-Phosphorylethanolamine (CAS 1071-23-4) is an endogenous phospholipid metabolite involved in the biosynthesis of phosphatidylethanolamine a major component of the cytoplasmic leaflet of cellular membranes It acts as a precursor in the CDP-ethanolamine pathway contributing to membrane integrity and dynamic cellular functions such as membrane fusion autophagy and apoptosis Research indicates that reduced levels of O-phosphorylethanolamine are associated with increased neuronal loss in neurodegenerative disorders including Alzheimer s Huntington s and Parkinson s diseases This molecule is therefore of interest in studies of phospholipid metabolism and neurodegeneration