Learn More
AdipoGen O-Phosphorylethanolamine
SDP
Supplier:  AdipoGen CDXC0021G025
Encompass_Preferred
Chemical. CAS 1071-23-4. Formula C2H8NO4P. MW 141.06. Synthetic. Phosphorylethanolamine is an ethanolamine derivative that is used in the synthesis of two different categories of phospholipids, glycerophospholipid and sphingophospholipid. It is used as an artificial AP-substrate in a TAMRA/TAMRA fluorescence-quenching assay. Newly discovered „phosphoethanolamine cellulose“ has been shown to regulate biofilm matrix architecture and function and might be important understanding bacterial biofilms and for the generation of new cellulosic materials. Synthetic phosphorylethanolamine PHO-S has recently demonstrated cytotoxic activity on melanoma tumor cells associated with apoptotic effects, without any effect on normal cells, as such human normal endothelial, fibroblasts and lymphocytes cells. It has shown to have in vitro and in vivo anti-leukemia effects.
Catalog No. 50-256-1543