Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Phosphorylethanolamine, 98%

Catalog No. AC367421000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC367421000 100 g Glass Bottle
AC367420050 5 g Glass Bottle
AC367420250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC367421000 Supplier Thermo Scientific Chemicals Supplier No. 367421000
Only null left
Add to Cart

CAS: 1071-23-4 | C2H8NO4P | 141.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-23-4
Molecular Formula C2H8NO4P
Molecular Weight (g/mol) 141.06
MDL Number MFCD00008178
InChI Key SUHOOTKUPISOBE-UHFFFAOYSA-N
Synonym o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
PubChem CID 1015
ChEBI CHEBI:17553
IUPAC Name 2-aminoethyl dihydrogen phosphate
SMILES C(COP(=O)(O)O)N

Specifications

Melting Point 241.0°C to 243.0°C
Assay Percent Range 97.5% min. (Titration with HClO4)
Packaging Glass Bottle
Linear Formula H2NCH2CH2OP(O)(OH)2
Quantity 100 g
Beilstein 04,IV,1415
Solubility Information Solubility in water: soluble.
Formula Weight 141.06
Percent Purity 98%
Chemical Name or Material O-Phosphorylethanolamine, 98%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
EINECSNumber 213-988-5
Recommended Storage Refrigerator +4°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.