Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-phthalaldehyde, 99.1%, MP Biomedicals™

Catalog No. ICN10264801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10264801 1 g
ICN10264805 5 g
ICN10264825 25 g
ICN10264880 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10264801 Supplier MP Biomedicals, Inc Supplier No. 0210264801
Add to Cart
Add to Cart

Chemical Identifiers

CAS 643-79-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00003335
InChI Key ZWLUXSQADUDCSB-UHFFFAOYSA-N
Synonym o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde
PubChem CID 4807
ChEBI CHEBI:70851
SMILES O=CC1=CC=CC=C1C=O

Specifications

Color Yellow
Assay Percent Range ≥98%
Quantity 1 g
IUPAC Name benzene-1,2-dicarbaldehyde
Formula Weight 134.13
Percent Purity 99.1%
Physical Form Crystals
Chemical Name or Material O-phthalaldehyde
Recommended Storage Store at +4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.