Learn More
Medchemexpress LLC O-phthalaldehyde-d6 | 68234-47-9 | MFCD30720408 | >95.0% | 140.17 | C8D6O2 | 5mg
SDP
Catalog No. 5000269745
Encompass_Preferred
Phthalaldehyde-d6 is the deuterium labeled Phthalaldehyde (HY-W012669) Phthalaldehyde is a biochemical assay reagent which modifies the amino acid and measure the derivative through HPLC Phthalaldehyde forms a fluorescent compound with -amino group[1][2]
Catalog No. 50-002-69745 Supplier Medchemexpress LLC Supplier No. HYW7384175MG
Phthalaldehyde-d6 is the deuterium labeled Phthalaldehyde (HY-W012669) Phthalaldehyde is a biochemical assay reagent which modifies the amino acid and measure the derivative through HPLC Phthalaldehyde forms a fluorescent compound with -amino group[1][2]