Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phthalaldehyde, MP Biomedicals™

Catalog No. MP210264810 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210264810 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210264810 Supplier MP Biomedicals, Inc Supplier No. 0210264810
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for precolumn derivatization of amino acids and other biochemical applications. o-Phthalaldehyde, MP Biomedicals is a dialdehyde, consisting of two formyl (CHO) groups attached to adjacent carbon centers on a benzene ring.

o-Phthalaldehyde is used for precolumn derivatization of amino acids for HPLC separation and for flow cytometric measurements of protein thiol groups. Used for fluorometric determination of histamine, histidine and other amino acids. Also used for cholesterol assay in the picomole range.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 643-79-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00003335
InChI Key ZWLUXSQADUDCSB-UHFFFAOYSA-N
Synonym o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde
PubChem CID 4807
ChEBI CHEBI:70851
IUPAC Name benzene-1,2-dicarbaldehyde
SMILES O=CC1=CC=CC=C1C=O

Specifications

Melting Point 53°C to 58°C
Quantity 10 g
Solubility Information Soluble in acetic acid (40mg/mL) or methanol (21mg/mL, clear solution); slightly soluble in water (5mg/mL).
Formula Weight 134.13
Percent Purity ≥98%
Physical Form Crystals
Recommended Storage Store at 4°C, protect from light, under nitrogen.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.