Learn More
AdipoGen o-Phthaldialdehyde
SDP
Supplier:  AdipoGen CDXO0008G025
Encompass_Preferred
Chemical. CAS 643-79-8. Formula C8H6O2. MW 134.13. Synthetic Flow cytometry measurement tool. Fluorescence probe for glutathione and glutathione disulfide levels, primary amines, peptides and biological thiols. Reacts with primary amines to produce a product that emits a highly fluorescent blue colour. Spectral data lambdaex 334 nm lambdaem 455 nm Thiol Adduct. Used for the quick visualization of histamine, characterized by the appearance of a yellow stain. Used for precolumn derivatization of amino acids for HPLC separation. High-level disinfectant/sterilant for endoscopes, thermometers, rubber and plastic equipment, and medical/surgical instruments that cannot be sterilized by heat, based on its antimicrobial efficacy and low inhalation toxicity.
Catalog No. 50-256-0349