Learn More
Medchemexpress LLC O-phthalimide-C5-acid | 4443-26-9 | MFCD00233499 | 98.0% | 261.27 g·mol^-1 | C14H15NO4 | 500 MG
SDP
Catalog No. 5000300170
Encompass_Preferred
O-phthalimide-C5-acid is a purified hapten (6-N-phthalimidoy hexanoic acid derivative) with a terminal carboxyl group supplied as a powder for research use. It is designed to be conjugated to carrier proteins for antigen design and biochemical assay development, providing a spacer arm bearing a phthalimide moiety.
  • Has terminal carboxyl group for conjugation to protein amines.
  • Functions as a hapten for antigen design and antibody generation.
  • Provides a phthalimide-containing spacer arm to present epitopes.
  • High purity (98.0%) suitable for biochemical assays.
  • Molecular weight 261.27 and formula C14H15NO4.
  • Available in multiple sizes for flexible experimental use.
  • For research use only.
Catalog No. 50-003-00170 Supplier Medchemexpress LLC Supplier No. HYW028210500MG
O-phthalimide-C5-acid is a purified hapten (6-N-phthalimidoy hexanoic acid derivative) with a terminal carboxyl group supplied as a powder for research use. It is designed to be conjugated to carrier proteins for antigen design and biochemical assay development, providing a spacer arm bearing a phthalimide moiety.
  • Has terminal carboxyl group for conjugation to protein amines.
  • Functions as a hapten for antigen design and antibody generation.
  • Provides a phthalimide-containing spacer arm to present epitopes.
  • High purity (98.0%) suitable for biochemical assays.
  • Molecular weight 261.27 and formula C14H15NO4.
  • Available in multiple sizes for flexible experimental use.
  • For research use only.