Learn More
Medchemexpress LLC HY-15680 10mg , O-Propargyl-Puromycin CAS:1416561-90-4 Purity:>98%
SDP
Catalog No. 502259083
Encompass_Preferred
HY-15680 10mg O-Propargyl-Puromycin CAS: 1416561-90-4 O-Propargyl-Puromycin, an alkyne analog of puromycin, is a potent protein synthesis inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9083 Supplier Medchemexpress LLC Supplier No. HY1568010MG
HY-15680 10mg O-Propargyl-Puromycin CAS: 1416561-90-4 O-Propargyl-Puromycin, an alkyne analog of puromycin, is a potent protein synthesis inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.