Learn More
Medchemexpress LLC O-Propargyl-Puromycin | 1416561-90-4 | 98.4% | 495.53 | 50 MG
SDP
Catalog No. 5000364269
Encompass_Preferred
O-Propargyl-Puromycin is a potent protein synthesis inhibitor and an alkyne analog of puromycin. It functions as a click chemistry reagent, featuring an Alkyne group that can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Protein synthesis inhibition: It effectively inhibits protein synthesis in both reticulocyte lysates and cultured cells.
  • Covalent conjugates: This compound forms covalent conjugates with nascent polypeptide chains.
  • Rapid turnover: These conjugates are rapidly turned over by the proteasome.
  • Visualization and capture: The covalent conjugates can be visualized or captured through copper(I)-catalyzed azide-alkyne cycloaddition.
  • Imaging and affinity purification: It can be utilized to image and affinity-purify nascent proteins within cells and animals.
Catalog No. 50-003-64269 Supplier Medchemexpress LLC Supplier No. 50MGHY15680
O-Propargyl-Puromycin is a potent protein synthesis inhibitor and an alkyne analog of puromycin. It functions as a click chemistry reagent, featuring an Alkyne group that can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Protein synthesis inhibition: It effectively inhibits protein synthesis in both reticulocyte lysates and cultured cells.
  • Covalent conjugates: This compound forms covalent conjugates with nascent polypeptide chains.
  • Rapid turnover: These conjugates are rapidly turned over by the proteasome.
  • Visualization and capture: The covalent conjugates can be visualized or captured through copper(I)-catalyzed azide-alkyne cycloaddition.
  • Imaging and affinity purification: It can be utilized to image and affinity-purify nascent proteins within cells and animals.