Learn More
TARGETMOL CHEMICALS INC O-Propargyl-Puromycin 1MG
SDP
Catalog No. 502879899
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. O-Propargyl-Puromycin (OP-puro) is a potent protein synthesis inhibitor, a puromycin acetylene analog. Purity 99.7%
Catalog No. 50-287-9899 Supplier TARGETMOL CHEMICALS INC Supplier No. T122841MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. O-Propargyl-Puromycin (OP-puro) is a potent protein synthesis inhibitor, a puromycin acetylene analog. Purity 99.7%