Learn More
Cayman Chemical O-Propargyl-puromycIn 1mg
SDP
Catalog No. 502971570
Encompass_Preferred
A clickable form of puromycin; inhibits protein synthesis in rabbit reticulocyte lysates and HEK293T cells from 1-25 µM; incorporated into nascent polypeptide chains where it forms covalent adducts and terminates chain elongation on the ribosome; has been used in combination with fluorescent azides to image nascent proteins in various cells
Catalog No. 50-297-1570 Supplier Cayman Chemical Supplier No. 151341 MG
A clickable form of puromycin; inhibits protein synthesis in rabbit reticulocyte lysates and HEK293T cells from 1-25 µM; incorporated into nascent polypeptide chains where it forms covalent adducts and terminates chain elongation on the ribosome; has been used in combination with fluorescent azides to image nascent proteins in various cells