Learn More
Cayman Chemical O-Propargyl-puromycIn 500ug
SDP
Catalog No. 502972792
Encompass_Preferred
A clickable form of puromycin; inhibits protein synthesis in rabbit reticulocyte lysates and HEK293T cells from 1-25 µM; incorporated into nascent polypeptide chains where it forms covalent adducts and terminates chain elongation on the ribosome; has been used in combination with fluorescent azides to image nascent proteins in various cells
Catalog No. 50-297-2792 Supplier Cayman Chemical Supplier No. 15134500 UG
A clickable form of puromycin; inhibits protein synthesis in rabbit reticulocyte lysates and HEK293T cells from 1-25 µM; incorporated into nascent polypeptide chains where it forms covalent adducts and terminates chain elongation on the ribosome; has been used in combination with fluorescent azides to image nascent proteins in various cells