Learn More
Description
- Length: 9 in.
- Serrated cutting blades provide strong grip and prevent tissue or bone slippage
- O.R. Grade construction with anti-glare satin finish ideal for operative lighting
- Designed for precise cutting of cartilage, small bones, and dense tissue
- Robust, high-leverage handle allows powerful cutting with reduced hand strain
- Made from premium surgical stainless steel for durability and corrosion resistance
- Autoclavable and reusable, suitable for repeated use in surgical settings
- Appropriate for orthopedic surgery, trauma procedures, autopsy, and postmortem applications
Specifications
Specifications
| Material | Stainless Steel |
| Length (English) | 9 in. |
| Length (Metric) | 22.9 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.