Learn More
Waters Corp Waters O-Ring, Needle Seal, Kalrez, 002
Catalog No. NC1839103 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This O-Ring serves as a replacement needle wash seal, crucial for maintaining the integrity of various Waters chromatography systems.
  • Functions as a replacement needle wash seal
  • Constructed from Kalrez material
  • Compatible with ACQUITY and NanoACQUITY Fixed Loop (FL) Sample Managers, UPC2, and UPLC systems
  • Recommended for annual replacement
Catalog No. NC1839103 Supplier Waters Corp Supplier No. 700002572
This O-Ring serves as a replacement needle wash seal, crucial for maintaining the integrity of various Waters chromatography systems.
  • Functions as a replacement needle wash seal
  • Constructed from Kalrez material
  • Compatible with ACQUITY and NanoACQUITY Fixed Loop (FL) Sample Managers, UPC2, and UPLC systems
  • Recommended for annual replacement