Learn More
Chemglass Life Sciences Chemglass O-Ring Simriz 486 FFKM, AS568-006 (White, USP Class VI)
SDP
Catalog No. 502743537
Encompass_Preferred
Simriz 486 FFKM perfluoroelastomer o-ring in AS568 size #006, designed for high-purity laboratory and semiconductor applications. The white, 75 Shore A material provides broad chemical resistance and stability at elevated temperatures, and is USP Class VI certified.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • White color for contamination-sensitive environments.
  • 75 Shore A durometer for resilient sealing performance.
  • USP Class VI certified for pharmaceutical and biotech use.
  • Available in AS568 standard sizes for direct replacement.
  • Suitable for vacuum traps, o-ring joints, and laboratory glassware seals.
Catalog No. 50-274-3537 Supplier Chemglass Life Sciences Supplier No. CG309Z006
Simriz 486 FFKM perfluoroelastomer o-ring in AS568 size #006, designed for high-purity laboratory and semiconductor applications. The white, 75 Shore A material provides broad chemical resistance and stability at elevated temperatures, and is USP Class VI certified.
  • Perfluoroelastomer (FFKM) construction for broad chemical resistance.
  • White color for contamination-sensitive environments.
  • 75 Shore A durometer for resilient sealing performance.
  • USP Class VI certified for pharmaceutical and biotech use.
  • Available in AS568 standard sizes for direct replacement.
  • Suitable for vacuum traps, o-ring joints, and laboratory glassware seals.