Learn More
Chemglass Life Sciences Chemglass O-Ring #007 - Simriz 485 FFKM perfluoro, black (CG-309-B-007)
SDP
Catalog No. 502743605
Encompass_Preferred
Chemglass O-Ring #007 (CG-309-B-007) is manufactured from Simriz 485 FFKM perfluoroelastomer to provide broad chemical resistance and high temperature performance for laboratory sealing applications. It serves as a replacement seal for compatible glassware, fittings, and laboratory equipment.
  • Perfluoroelastomer material for broad chemical resistance.
  • Operating temperature -7°C to +230°C.
  • Hardness 75 Shore A.
  • High tensile strength (2690 psi) for durability.
  • Black color for visual identification.
Catalog No. 50-274-3605 Supplier Chemglass Life Sciences Supplier No. CG309B007
Chemglass O-Ring #007 (CG-309-B-007) is manufactured from Simriz 485 FFKM perfluoroelastomer to provide broad chemical resistance and high temperature performance for laboratory sealing applications. It serves as a replacement seal for compatible glassware, fittings, and laboratory equipment.
  • Perfluoroelastomer material for broad chemical resistance.
  • Operating temperature -7°C to +230°C.
  • Hardness 75 Shore A.
  • High tensile strength (2690 psi) for durability.
  • Black color for visual identification.