Learn More
Chemglass Life Sciences Chemglass O-Ring #007 - Simriz 486 FFKM (Perfluoro), White, USP Class VI - Sealing part for laboratory fittings and components
SDP
Catalog No. 502743536
Encompass_Preferred
O-ring made from a high-purity perfluoroelastomer compound designed for laboratory sealing applications. Size #007, white color, and USP Class VI certification make this part suitable for aggressive dry process chemistries and reactive plasma environments.
  • High-purity perfluoroelastomer material.
  • White color for visual inspection.
  • Certified to USP Class VI.
  • Resistant to a wide range of chemicals and solvents.
  • Heat resistant to 220°C.
  • Suitable for aggressive dry process chemistries and reactive plasma applications.
  • Manufactured in ISO-certified clean-room facilities.
Catalog No. 50-274-3536 Supplier Chemglass Life Sciences Supplier No. CG309Z007
O-ring made from a high-purity perfluoroelastomer compound designed for laboratory sealing applications. Size #007, white color, and USP Class VI certification make this part suitable for aggressive dry process chemistries and reactive plasma environments.
  • High-purity perfluoroelastomer material.
  • White color for visual inspection.
  • Certified to USP Class VI.
  • Resistant to a wide range of chemicals and solvents.
  • Heat resistant to 220°C.
  • Suitable for aggressive dry process chemistries and reactive plasma applications.
  • Manufactured in ISO-certified clean-room facilities.