Learn More
Chemglass Life Sciences Chemglass O-ring #008 - Simriz 486 FFKM perfluoroelastomer, white, USP Class VI (for vacuum traps and O-ring joints)
SDP
Catalog No. 502743535
Encompass_Preferred
This white FFKM O-ring is a high-purity perfluoroelastomer seal designed for laboratory glassware applications. It delivers broad chemical resistance and stable performance at elevated temperatures, and is certified to USP Class VI for biocompatibility.
  • High chemical resistance to a broad range of solvents and reagents.
  • White, high-purity perfluoroelastomer material.
  • Certified to USP Class VI for biocompatibility.
  • Numbered size: #008, suitable for small glassware joints.
  • Intended for use with vacuum traps and O-ring joints.
Catalog No. 50-274-3535 Supplier Chemglass Life Sciences Supplier No. CG309Z008
This white FFKM O-ring is a high-purity perfluoroelastomer seal designed for laboratory glassware applications. It delivers broad chemical resistance and stable performance at elevated temperatures, and is certified to USP Class VI for biocompatibility.
  • High chemical resistance to a broad range of solvents and reagents.
  • White, high-purity perfluoroelastomer material.
  • Certified to USP Class VI for biocompatibility.
  • Numbered size: #008, suitable for small glassware joints.
  • Intended for use with vacuum traps and O-ring joints.