Learn More
Perkin Elmer US LLC Kalrez Small Seal O-Ring - High Temperature, Size 009
Catalog No. 509052152
Encompass_Preferred
Designed for use in the TurboMatrix Upper Seal Assembly and at high temperatures (above 225°C).
Catalog No. 50-905-2152 Supplier Perkin Elmer US LLC Supplier No. M0415360
Designed for use in the TurboMatrix Upper Seal Assembly and at high temperatures (above 225°C).