Learn More
Chemglass Life Sciences Chemglass O-Ring CG-309-Z-009 (Simriz 486 FFKM, AS568-009)
SDP
Catalog No. 502743534
Encompass_Preferred
Chemglass O-ring CG-309-Z-009 is a white perfluoroelastomer (FFKM) sealing ring manufactured from Simriz 486 compound for high-purity laboratory and process applications. It is USP Class VI certified and designed to provide broad chemical resistance and elevated temperature stability in pharmaceutical, semiconductor, and biotech equipment. The ring is supplied in AS568 standard sizes, including the applicable AS568-009 size.
  • Perfluoroelastomer construction for broad chemical compatibility.
  • USP Class VI certified for biocompatibility.
  • White color suited for contamination-sensitive applications.
  • Operating temperature range of -7°C to +230°C.
  • Hardness of 75 Shore A for durable sealing performance.
  • Tensile strength approximately 2,620 psi.
  • Supplied in a wide range of AS568 standard sizes for compatibility.
Catalog No. 50-274-3534 Supplier Chemglass Life Sciences Supplier No. CG309Z009
Chemglass O-ring CG-309-Z-009 is a white perfluoroelastomer (FFKM) sealing ring manufactured from Simriz 486 compound for high-purity laboratory and process applications. It is USP Class VI certified and designed to provide broad chemical resistance and elevated temperature stability in pharmaceutical, semiconductor, and biotech equipment. The ring is supplied in AS568 standard sizes, including the applicable AS568-009 size.
  • Perfluoroelastomer construction for broad chemical compatibility.
  • USP Class VI certified for biocompatibility.
  • White color suited for contamination-sensitive applications.
  • Operating temperature range of -7°C to +230°C.
  • Hardness of 75 Shore A for durable sealing performance.
  • Tensile strength approximately 2,620 psi.
  • Supplied in a wide range of AS568 standard sizes for compatibility.