Learn More
Chemglass Life Sciences Chemglass O-ring #010 - Simriz 485 FFKM perfluoroelastomer, black; replacement seal for laboratory glassware and fittings
SDP
Catalog No. 502743604
Encompass_Preferred
Perfluoroelastomer O-ring, size #010, designed as a replacement seal for laboratory glassware assemblies and fittings that accept #010 O-rings.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Black color.
  • Size: #010.
  • Hardness: Shore A 75.
  • Temperature range: -7°C to +230°C.
  • Tensile strength: 2690 psi.
  • Suitable as a replacement seal for glassware fittings and assemblies.
Catalog No. 50-274-3604 Supplier Chemglass Life Sciences Supplier No. CG309B010
Perfluoroelastomer O-ring, size #010, designed as a replacement seal for laboratory glassware assemblies and fittings that accept #010 O-rings.
  • Perfluoroelastomer (FFKM) material for broad chemical resistance.
  • Black color.
  • Size: #010.
  • Hardness: Shore A 75.
  • Temperature range: -7°C to +230°C.
  • Tensile strength: 2690 psi.
  • Suitable as a replacement seal for glassware fittings and assemblies.