Learn More
Chemglass Life Sciences Chemglass O-Ring Simriz 486 FFKM Perfluoro, AS568-010, White, 75 Shore A
SDP
Catalog No. 502743533
Encompass_Preferred
Perfluoroelastomer O-ring manufactured from Simriz 486 FFKM in AS568 size #010. White, 75 Shore A hardness, and certified to USP Class VI, this O-ring is engineered for high-purity and aggressive chemical environments and elevated temperature sealing up to ~220°C.
  • High resistance to aggressive and reactive chemistries.
  • White color aids visual cleanliness and contamination control.
  • USP Class VI certified for biocompatibility and purity.
  • High temperature performance suitable for elevated process conditions.
  • 75 Shore A hardness provides resilient sealing and compression set resistance.
  • Supplied in standard AS568 sizes for interchangeability.
Catalog No. 50-274-3533 Supplier Chemglass Life Sciences Supplier No. CG309Z010
Perfluoroelastomer O-ring manufactured from Simriz 486 FFKM in AS568 size #010. White, 75 Shore A hardness, and certified to USP Class VI, this O-ring is engineered for high-purity and aggressive chemical environments and elevated temperature sealing up to ~220°C.
  • High resistance to aggressive and reactive chemistries.
  • White color aids visual cleanliness and contamination control.
  • USP Class VI certified for biocompatibility and purity.
  • High temperature performance suitable for elevated process conditions.
  • 75 Shore A hardness provides resilient sealing and compression set resistance.
  • Supplied in standard AS568 sizes for interchangeability.