Learn More
Perkin Elmer US LLC Kalrez Large Seal O-Ring - High Temperature, Size 011
Catalog No. 509052153
Encompass_Preferred
Designed for use in the TurboMatrix Upper Seal Assembly and at high temperatures (above 225°C).
Catalog No. 50-905-2153 Supplier Perkin Elmer US LLC Supplier No. M0415361
Designed for use in the TurboMatrix Upper Seal Assembly and at high temperatures (above 225°C).