Learn More
Chemglass Life Sciences Chemglass O-Ring CG-309-Z-011, Simriz 486 FFKM (AS568-011)
SDP
Catalog No. 502743532
Encompass_Preferred
A high-purity white perfluoroelastomer (FFKM) O-ring formulated from Simriz 486 for demanding chemical and temperature environments. Certified to USP Class VI, it is intended for pharmaceutical, semiconductor, and high-purity laboratory sealing applications.
  • High chemical resistance suitable for aggressive chemistries.
  • Temperature resistance up to 220°C.
  • White color aids contamination control in clean environments.
  • Certified to USP Class VI for biocompatibility.
  • Suitable for pharmaceutical, semiconductor, and biotech applications.
  • Available in AS568 standard sizes for common sealing uses.
Catalog No. 50-274-3532 Supplier Chemglass Life Sciences Supplier No. CG309Z011
A high-purity white perfluoroelastomer (FFKM) O-ring formulated from Simriz 486 for demanding chemical and temperature environments. Certified to USP Class VI, it is intended for pharmaceutical, semiconductor, and high-purity laboratory sealing applications.
  • High chemical resistance suitable for aggressive chemistries.
  • Temperature resistance up to 220°C.
  • White color aids contamination control in clean environments.
  • Certified to USP Class VI for biocompatibility.
  • Suitable for pharmaceutical, semiconductor, and biotech applications.
  • Available in AS568 standard sizes for common sealing uses.