Learn More
Chemglass Life Sciences Simriz 486 FFKM O-Ring AS568-012 - 0.364 in ID x 0.070 in cross section, white (for AS568 groove seals and laboratory/process equipment)
SDP
Catalog No. 502743531
Encompass_Preferred
Perfluoroelastomer (FFKM) O-ring made from the Simriz 486 (SZ486) compound in AS568-012 size. Designed for demanding pharmaceutical, semiconductor, and biotech applications that require broad chemical compatibility, thermal stability, and low particulate behavior.
  • Perfluoroelastomer material provides exceptional chemical resistance and thermal stability.
  • White, low-particulate compound suitable for cleanroom and pharmaceutical environments.
  • 75 durometer offers durable sealing with low compression set for long service life.
  • Standard AS568 size enables interchangeability with common groove standards.
  • Available in single-piece quantities for maintenance, repair, and spare parts.
Catalog No. 50-274-3531 Supplier Chemglass Life Sciences Supplier No. CG309Z012
Perfluoroelastomer (FFKM) O-ring made from the Simriz 486 (SZ486) compound in AS568-012 size. Designed for demanding pharmaceutical, semiconductor, and biotech applications that require broad chemical compatibility, thermal stability, and low particulate behavior.
  • Perfluoroelastomer material provides exceptional chemical resistance and thermal stability.
  • White, low-particulate compound suitable for cleanroom and pharmaceutical environments.
  • 75 durometer offers durable sealing with low compression set for long service life.
  • Standard AS568 size enables interchangeability with common groove standards.
  • Available in single-piece quantities for maintenance, repair, and spare parts.